Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD94 Mouse anti-Human, FITC, Clone: HP-3D9, BD

Catalog No. BDB555888 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555888 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555888 Supplier BD Biosciences Supplier No. 555888
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 70kDa disulfide-linked dimer, also known as Kp43, expressed on natural killer (NK) cells. It is also expressed on γ/δ TCR+ T lymphocytes and on some CD8+CD56+ α/β TCR+ clones. Reports demonstrate that HP-3D9 markedly inhibits cytolytic activity of polyclonally-activated NK cells. CD94 (Kp43) plays a role in regulation of the function of NK cell activation and adhesion

Host Species: Mouse
Clone: HP-3D9
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD94
Applications Flow Cytometry
Classification Monoclonal
Clone HP-3D9
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NK Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.