Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD95 Hamster anti-Mouse, Brilliant Violet 421, Clone: Jo2, BD

Catalog No. BDB562633 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562633 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562633 Supplier BD Biosciences Supplier No. 562633
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hamster Monoclonal Antibody

Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Antibody was conjugated with Brilliant Violet 421 under optimum conditions, and unconjugated antibody and free Brilliant Violet 421 were removed. Monoclonal antibodies recognizing Fas such as Jo2 have cytolytic activity on cells expressing Fas.

Host Species: Hamster
Clone: Jo2
Isotype: Ar Ham IgG2, λ2
Species Reactivity: Mouse
Immunogen: WR19L mouse lymphoma cells transformed with recombinant mouse Fas

  • Jo2 antibody shows cytolytic activity against cell lines expressing mouse Fas by inducing apoptosis
  • Jo2 mAb has been reported to immunoprecipitate mouse Fas as 45 kDa band from W4 cells (WR19L mouse lymphoma cells transformed with mouse Fas)

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD95
Applications Flow Cytometry
Classification Monoclonal
Clone Jo2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Apo-1; Apt1; Fas; FASLG receptor; lpr; TNFR6; Tnfrsf6; TNR6
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Hamster
Immunogen WR19L mouse lymphoma cells transformed with recombinant mouse Fas
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.