Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD98 Mouse anti-Human, Unlabeled, Clone: UM7F8, BD

Catalog No. BDB556074 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556074 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556074 Supplier BD Biosciences Supplier No. 556074
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 125kDa, disulfide-linked heterodimer composed of a glycosylated heavy chain, type II membrane glycoprotein and a non-glycosylated light chain. CD98 is broadly expressed on peripheral blood lymphocytes, monocytes and granulocytes (low). It is also expressed on non-hemtopoietic cells. Reports suggest that CD98 may be involved in the regulation of cellular activation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development only or reported in the literature.

Host Species: Mouse
Clone: UM7F8
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD98
Applications Flow Cytometry
Classification Monoclonal
Clone UM7F8
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human T-leukemic and Thymic Cell Lines
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.