Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC42 Mouse, Unlabeled, Clone: 44, BD

Catalog No. BDB610928 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610928 50 μg
BDB610929 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610928 Supplier BD Biosciences Supplier No. 610928
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rho family members are small GTP binding proteins that serve as molecular switches for a number of biological processes. They cycle between active GTP-bound and inactive GDP-bound states. CDC42 is a Rho family protein that was identified in membranes of human platelets and placenta. It is the homologue of CDC42Sc, which regulates initiation of bud-site assembly in Saccharomyces cerevisiae. CDC42 regulates the function of the mammalian actin cytoskeleton, allowing for efficient cytokinesis and cell morphogenesis. CDC42 and Rac1, a Ras-related GTPase, activate MEKK1, a JNK kinase kinase, which leads to the activation of several downstream components of the MAP kinase cascade leading to activation of PAK65, a CDC42- and Rac-binding protein. PAK65 interacts with CDC42/Rac1, mediates their interaction with MEKK1, and enhances MEKK1 catalytic activity. Chronic activation of CDC42 has been shown to induce malignant cellular transformation. Due to sequence homology among various small GTPase family members, potential cross-reactivity could be observed with this antibody. BD Pharmingen has not performed such cross-reactivity testing.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC42
Applications Western Blot
Classification Monoclonal
Clone 44
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CDC42 aa. 1-191
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.