Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Anti-Cdc42 Immunoblotting Kit

Catalog No. 17299 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Immunoblots/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-299 10 Immunoblots/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-299 Supplier MilliporeSigma™ Supplier No. 17299
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Anti-cdc42, (mouse monoclonal IgG1)
  • Immunoblot Blocking Reagent
  • Goat Anti-Mouse IgG, HRP conjugate

Detects Cdc42 when performing western blots

Anti-cdc42 is a mouse monoclonal IgG1 that recognizes cdc42 from human, mouse, rat and dog
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blotting
Quantity 10 Immunoblots/Kit
Stability Stable for 1 year at -20°C from date of shipment
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.