Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk1/Cdc2 (pY15) Mouse, Unlabeled, Clone: 44, BD

Catalog No. p-3879074 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612306 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612306 Supplier BD Biosciences Supplier No. 612306
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Progression of the mammalian cell cycle is regulated by phosphorylation of many key proteins. Several classes of cyclins (A-E) act as regulatory subunits for cyclin-dependent kinases (cdks). Cdk1/Cdc2 (p34 [cdc2]) is the catalytic subunit of the maturation promoting factor (MPF), which includes the regulatory subunit cyclin B. During late S and G2 phase, cyclin B synthesis increases, allowing it to bind Cdc2. This begins the transition into M-phase of the mammalian cell cycle by initiating a series of phosphorylation and dephosphorylation events that lead to activation of the Cdc2/cyclin B complex. After binding to cyclin B, cdc2 is phosphorylated on Thr-14, by Myt1, and Tyr-15, by wee1 or mik1, yielding an inactive pre-MPF complex during G2 phase. Phosphorylation of cdc2 on Thr-161 is performed by a cdk7/cyclin H complex and is necessary for activation of the cdc2 complex. Dephosphorylation of Thr-14 and Tyr-15 by CDC25 occurs at the end of G2 phase and completes activation of the cdc2/Cyclin B complex and facilitates entry into mitosis. During mitosis, cyclin B is targeted for degradation and Cdc2 becomes inactive again.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk1/Cdc2 (pY15)
Applications Western Blot
Classification Monoclonal
Clone 44
Concentration 250μg/mL
Conjugate Unconjugated
Description p34 [cdc2]
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human Cdc2 (pY15) Peptide
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.