Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK5RAP2 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 061398MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-139-8MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-139-8MI Supplier MilliporeSigma Supplier No. 061398
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-CDK5RAP2 Antibody (rabbit polyclonal antibody) validated in western blot, immunocytochemistry to detect CDK5RAP2 also known as CDK5 regulatory subunit-associated protein 2, CDK5 activator-binding protein C48, Centrosome-associated protein 215.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK5RAP2
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols CDK5RAP2
Host Species Rabbit
Immunogen MBP-tagged recombinant protein corresponding to human CDK5RAP2.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001011649
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.