Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX-2 Mouse anti-Human, PE, Clone: M39-71, BD

Catalog No. BDB563428 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563428 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563428 Supplier BD Biosciences Supplier No. 563428
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The M39-711 monoclonal antibody reacts with caudal-type homeobox transcription factor 2 (CDX-2). In embryonic development, CDX-2 is first expressed in the outer layer of the morula and later in the trophoblast, but not in embryonic stem cells. Along with Oct3/4, it is involved in the segregation of the trophoectoderm lineage from the inner cell mass. In the adult, CDX-2 expression is limited to the mucosa of the small and large intestines, where it regulates the synthesis of several intestine-specific proteins. Loss of CDX-2 expression is observed in some colorectal carcinomas, while gain of CDX-2 expression has been observed in gastric tumors and acute myeloid leukemias.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CDX-2
Applications Flow Cytometry
Classification Monoclonal
Clone M39-71
Conjugate PE
Description CDX2, CDX3, CDX-3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CDX-2 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.