Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramphenicol Acetyltransferase Mouse anti-Vertebrates, E. coli, Clone: CAT-1, MilliporeSigma™

Catalog No. MAB3678MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3678MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3678MI Supplier MilliporeSigma Supplier No. MAB3678
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Chloramphenicol Acetyltransferase using this Anti-Chloramphenicol Acetyltransferase Antibody, clone CAT-1 validated for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chloramphenicol Acetyltransferase
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone CAT-1
Conjugate Unconjugated
Formulation Ascites fluid. Liquid. Contains no preservative.
Host Species Mouse
Immunogen Chloramphenicol acetyl transferase
Quantity 100 μL
Primary or Secondary Primary
Target Species E. coli, Vertebrates
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.