Learn More
Immunology Consultants Affinity Purified Goat anti-Hamster (CHO) Nucleobindin-2 Antibody, 0.5 mg
SDP
Supplier:  Immunology Consultants GNUC65ALY
Encompass_Preferred
Goats were immunized with CHO nucleobindin-2 (NUC) of recombinant origin and antiserum were collected. The antibody was immunoaffinity purified off an antigen containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with CHO NUC of recombinant origin and NUC secreted by a null CHO cell line as determined by ELISA and western blot. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. NC2734221
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.