Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choline Acetyltransferase Goat anti-Mouse, Rat, Human, Chicken, Avian, Guinea Pig, Opossum, Polyclonal, MilliporeSigma™

Catalog No. AB144P200UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB144P200UL 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB144P200UL Supplier MilliporeSigma Supplier No. AB144P200UL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

This Anti-Choline Acetyltransferase (ChAT) Antibody is validated for use in immunocytochemistry, immunohistochemistry, western blotting for the detection of Choline Acetyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Choline Acetyltransferase
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified goat polyclonal in buffer containing PBS containing 5mg/mL BSA and 0.2% sodium azide.
Gene Symbols CHAT; ChAT; CHOACTase; CMS1A; CMS1A2
Host Species Goat
Immunogen Human placental enzyme.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_065574
Target Species Avian, Chicken, Guinea Pig, Human, Mouse, Opossum, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.