Learn More
MBL International Corporation Anti-cIAP 1and 2
SDP
Supplier:  MBL International Corporation CYP1041
Encompass_Preferred
Clonality:Polyclonal; Application:WB; Conjugate:Unlabeled; Species Reactivity:Human|Mouse|Rat; Size:100µg; Target:cIAP 1 and 2; The cellular inhibitor of apoptosis 1 and 2 (c-IAP1 and c-IAP2) proteins were identified based on their homology to a family of viral and cellular IAP proteins known to bind to caspases and inhibit their activity .This polyclonal antibody detects endogenous both c-IAP1 and c-IAP2 by western blotting.Anti-cIAP 1and 2 Polyclonal Antibody.
Catalog No. 50-304-564
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.