Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLIP-115 Mouse, Unlabeled, Clone: 14, BD

Catalog No. p-3970456 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611142 50 μg
BDB611143 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611142 Supplier BD Biosciences Supplier No. 611142

Mouse Monoclonal Antibody

Microtubules, integral components of the cytoskeleton, are essential for establishment of cell polarity, membrane structure organization, and organelle translocation. Cell polarity is accomplished through the linkage of microtubules to other cytoskeletal proteins by microtubule associated proteins (MAPs). Likewise, the process of organelle translocation requires accessory proteins that link the organelles to microtubules. These proteins are known as CLIPs (cytoplasmic linker proteins) and belong to a novel family of microtubule binding proteins that are characterized by the presence of N-terminal microtubule binding (MTB) domains. CLIP-115 was discovered due to its high degree of homology to CLIP-170 (restin), which links endosomes to microtubules. Like CLIP-170, CLIP-115 contains two MTB domains and a long coiled-coil region. CLIP-115 has been reported to be found primarily in the brain where it associates with microtubules and the dendritic lamellar body (DLB). DLBs have been reported to be organelles that are present exclusively in the bulbous dendritic appendages of neurons. It is thought that CLIP-115 is a protein constituent of DLBs and is responsible for determining their structure and cytoplasmic location.

Host Species: Mouse
Clone: 14
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat CLIP-115 aa. 801-917
Formula Weight [Chemical]: 115kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIP-115
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 14
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Rat CLIP-115 aa. 801-917
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.