Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB756PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB756PMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB756PMI Supplier MilliporeSigma Supplier No. AB756P
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Collagen Antibody, Type IV is validated for use in ELISA, immunofluoresence, immunohistochemistry (paraffin) for the detection of Collagen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen
Applications ELISA, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.01M phosphate, 0.09M NaCl, pH 7.2 with no preservatives.
Host Species Rabbit
Immunogen Collagen Type IV extracted and purified from mouse tumor tissues.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.