Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortisol Mouse anti-Human, Clone: F4P1A3, MilliporeSigma™

Catalog No. MAB1243MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1243MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1243MI Supplier MilliporeSigma Supplier No. MAB1243
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cortisol Antibody, 3-CMO, clone F4P1A3 is an antibody against Cortisol for use in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cortisol
Applications ELISA
Classification Monoclonal
Clone F4P1A3
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS, pH 7.4 with 50% glycerol and 0.1% sodium azide.
Host Species Mouse
Immunogen Cortisol-3-CMO-BSA
Quantity 500 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.