Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREBL1, Mouse, Clone: 4D10, Abnova™

Catalog No. 89021160 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-160 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-160 Supplier Abnova Corporation Supplier No. H00001388M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CREBL1.

The protein encoded by this gene is a transcription factor in the unfolded protein response (UPR) pathway during ER stress. Either as a homodimer or as a heterodimer with ATF6-alpha, the encoded protein binds to the ER stress response element, interacting with nuclear transcription factor Y to activate UPR target genes. The protein is normally found in the membrane of the endoplasmic reticulum; however, under ER stress, the N-terminal cytoplasmic domain is cleaved from the rest of the protein and translocates to the nucleus. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: AELMLLSEIADPTRFFTDNLLSPEDWGLQNSTLYSGLDEVAEEQTQLFRCPEQDVPFDGSSLDVGMDVSPSEPPWELLPIFPDLQVK

Specifications

Antigen CREBL1
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4D10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CREBL1.
Formulation PBS with no preservative; pH 7.4
Gene ATF6B
Gene Accession No. NM_004381
Gene Alias CREB-RP/CREBL1/FLJ10066/G13
Gene Symbols ATF6B
Host Species Mouse
Immunogen CREBL1 (NP_004372.3, 2 a.a. ∼ 88 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1388
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.