Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Cryptococcus neoformans, Clone: CSFi, Epredia™ LabVision

Catalog No. p-3037657 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS622P0 100 μL 200μg/mL
MS622P1 500 μL 200μg/mL
MS622P 1 mL 200μg/mL
MS622P1ABX 100 μL 1mg/mL
MS622PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS622P0 Supplier Epredia Supplier No. MS622P0

Mouse Monoclonal Antibody

The most important opportunistic fungal pathogens include Cryptococcus neoformans, Candida and Aspergillus species, and the fungi that cause mucormycosis. C. neoformans resides and survives in an acidic phagolysosome. Disseminated cryptococcosis is accompanied by cryptococcal polysaccharides in the serum and the lack of cellular infiltrates in infected tissues. C. neoformans stimulates fetal blood lymphocytes to proliferate and stimulates proliferation of CD45RA+ cells from adults, indicating that it stimulates naive T cells.

Host Species: Mouse
Clone: CSFi
Isotype: IgG2b
Cross Reactivity: All strains of C neoformans
Epitope: Carbohydrate moiety occuring on one or two glycoproteins.
Immunogen: Formalin-treated encapsulated clinical isolate, CSF-1.
Molecular Weight: ∽50kDa and ∽90kDa
Positive Control: Brain tissue infected with C. neoformans.
Cellular Localization: Cell wall.

Ab-1 binds to glycoproteins with MWs of ∽50kDa and ∽90kDa found in the yeast cell wall, on its surface, and secreted into the culture medium. Binding of Ab-1 to cell-associated antigens opsonizes the yeast and enhances their phagocytosis. NeoMarkers' Ab-1 identifies all (encapsulated and acapsular) strains of C. neoformans and is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Electron Microscopy, Immunofluorescence, Immunohistochemistry, Western Blotting

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen Cryptococcus neoformans Ab-1
Classification Monoclonal
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Formalin-treated encapsulated clinical isolate, CSF-1.
Quantity 100 μL
Primary or Secondary Primary
Form Purified
Isotype IgG2b
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.