Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUL-3 Mouse, Unlabeled, Clone: 3, BD

Catalog No. BDB611848 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611848 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611848 Supplier BD Biosciences Supplier No. 611848
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The cullins are a family of proteins that are integral to cell cycle regulation. Members of this family include human Cul proteins, C.elegans Ce-Cul-1, and S. cerevisiae Cdc53. Proper control of cell cycle progression is essential for the prevention of tumorigenesis. Mutant forms of both Ce-Cul-1 and Cdc53 have been implicated in the oncogenic process. The human Cul family of proteins, Cul-1, -2, -3, -4A, -4B, and -5, have also been implicated in oncogenic processes. Abnormal nuclear localization of Cul-2 is seen in a rare hereditary condition known as VHL (von Hippel-Lindau) syndrome. Patients with VHL syndrome possess a mutant VHL gene and develop cancers such as retinal anginomas, CNS hemangioblastomas, and renal cell carcinomas. Cul-3 is widely expressed in normal cells and tissues, and shows increased expression in colon cancer cells. Cul-3 can associate with cyclin E in mammalian cells, and overexpression of Cul-3 targets cyclin E for ubiquination, while deletion of Cul-3 in mice leads to increased cyclin E accumulation and abnormal regulation of S-phase. Thus, the Cul
family of proteins may have roles in cell cycle regulation and tumoriogenesis.

Host Species: Mouse
Clone: 3
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Human CUL-3 aa. 565-684

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CUL-3
Applications Western Blot
Classification Monoclonal
Clone 3
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CUL-3 aa. 565-684
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.