Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCR5, Alexa Fluor 700, Clone: 51505, R&D Systems™

Catalog No. p-4712940
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB190N025 25 Tests
FAB190N100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB190N025 Supplier R&D Systems Supplier No. FAB190N025
This item is not returnable. View return policy

Host Species: Mouse
Clone: 51505
Isotype: IgG2B
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CXCR5. Met1-Phe372

Applications:

Flow Cytometry

Specifications

Antigen CXCR5
Applications Flow Cytometry
Classification Monoclonal
Clone 51505
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Gene Accession No. P32302
Gene Alias BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15
Gene Symbols CXCR5
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR5, Met1-Phe372, Accession # P32302
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 643
Test Specificity Stains human CXCR5 transfectants but not the parental cell lines in flow cytometry. Does not cross-react with human CXCR2, CXCR3, or CXCR4 transfectants.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.