Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CXCR7/RDC-1, PE, Clone: 358426, R&D Systems
Manufacturer: R&D Systems FAB42271P025
Description
Host Species: Mouse
Clone: 358426
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CXCR7/RDC?1. Met1-Lys362 (Gly131Ser)
Applications:
Flow Cytometry
Specifications
CXCR7/RDC-1 | |
358426 | |
Flow Cytometry 10 uL/10^6 cells | |
AAA62370 | |
ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
Mouse | |
IgG2a | |
25 Tests | |
Primary | |
57007 | |
Human |
Flow Cytometry | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Purified | |
CXCR7 | |
NS0 mouse myeloma cell line transfected with human CXCR7/RDC-1, Met1-Lys362 (Gly131Ser), Accession # AAA62370 | |
Protein A or G purified from hybridoma culture supernatant | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
Monoclonal | |
Detects human CXCR7/RDC-1 in direct ELISAs. In flow cytometry, reacts specifically with five distinct human CXCR7 transfectants, but does not react with their respective parental lines or mouse CXCR7 transfectants. In flow cytometry, also reacts with monocytes expressing CXCR7, but does not react with MCF-7 cells which have been reported to have surface-expressing CXCR7 using clone 11G8. Due to the conflicting reports published, use of clone 358426 may result in an underestimation of CXCR7 expression on certain cell types. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title