Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B Mouse, Unlabeled, Clone: 18, BD

Catalog No. BDB610220 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610220 150 μg
BDB610219 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610220 Supplier BD Biosciences Supplier No. 610220
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The B-type cyclins serve as subunits of the major mitotic protein kinase in many different organisms. In mammals, Cyclin B is a subunit for the cdc2 (Cdk1) protein kinase and is synthesized during late S and G2 phases. Cyclin B associates with inactive cdc2 and facilitates phosphorylation of cdc2 at Thr-14 and Tyr-15. This phosphorylation maintains the inactive state until the end of G2. The inactive Cyclin B-cdc2 complex continues to accumulate in the cytoplasm until the CDC25 phosphatase dephosphorylates Thr-14 and Tyr-15, rendering the complex active at the G2/M boundary. This mitotic kinase complex remains active until the metaphase/anaphase transition when Cyclin B becomes degraded. This degradation process appears to be ubiquitin-dependent and is necessary for the cell to exit mitosis. Also, the conserved domain in B-type cyclins, known as the P box, is required for CDC25 activation.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin B
Applications Western Blot
Classification Monoclonal
Clone 18
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.