Learn More
Advanced ImmunoChemical, Inc. ANTI-CYSTATIN C MAB
Supplier:  Advanced ImmunoChemical, Inc. 2CYSC1 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs:Cyst10, Cyst13, Cyst16, Cyst18, Cyst23, Cyst28New MAbs: Cyst11, Cyst20, Cyst29Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with cystatin C purified from human urine.Specificity:Cystatin C.MAb Isotypes:IgG1 for MAbs Cyst11, Cyst13, Cyst16, Cyst18, Cyst20, Cyst23, and Cyst28 IgG2a for MAb Cyst29 IgG3 for MAb Cyst10Applications:Cystatin C immunoassay, human cystatin C sandwich immunoassay. Recommended pairs for sandwich immunoassay (capture-detection):Cyst28Cyst23 – Cyst13Recommended pairs with the best cross-reaction with animal sera (capture-detection):Cyst11-Cyst20 (dog serum)Cyst29-Cyst16 (cat serum)Cyst29-Cyst20 (cat serum)Cyst29-Cyst20 (horse serum)Cyst20-Cyst13 (horse serum)Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C
Catalog No. NC3566217