Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoglein Mouse, Unlabeled, Clone: 62, BD

Catalog No. p-3855489 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610273 50 μg
BDB610274 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610273 Supplier BD Biosciences Supplier No. 610273
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Desmoglein (Dsg1) is a transmembrane glycoprotein found in desmosome junctions. Desmosomes and adherens junctions are plasma membrane specializations involved in cell-cell adhesion. They serve as membrane anchorage domains for intermediate filaments and microfilaments, respectively. Actin microfilaments anchor at adherens junctions, which occur in many sizes, shapes, and positions. These junctions generally contain E- or N-Cadherin whose cytoplasmic region is associated with cytoplasmic proteins such as α- and β- catenin, vinculin, α-actinin, and radixin. Intermediate filaments (IFs) are anchored at desmosomes which contain desmosome-specific cadherin family members, known as desmocollin (Dsc) and desmoglein. These proteins associate with cytoplasmic proteins such as desmoplakin and plakoglobin (γ-Catenin). Plakoglobin is the only junctional protein found at both desmosomes and adherens junctions and, therefore, plays a crucial role in cell adhesion.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoglein
Applications Western Blot
Classification Monoclonal
Clone 62
Concentration 250μg/mL
Conjugate Unconjugated
Description Dsg1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Drosophila, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.