Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGCR6, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002807 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-807 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-807 Supplier Abnova Corporation Supplier No. H00008214B01P
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human DGCR6 protein.

DiGeorge syndrome, and more widely, the CATCH 22 syndrome, are associated with microdeletions in chromosomal region 22q11.2. The product of this gene shares homology with the Drosophila melanogaster gonadal protein, which participates in gonadal and germ cell development, and with the gamma-1 subunit of human laminin. This gene is a candidate for involvement in DiGeorge syndrome pathology and in schizophrenia. [provided by RefSeq

Sequence: MERYAGALEEVADGARQQERHYQLLSALQSLVKELPSSFQQRLSYTTLSDLALALLDGTVFEIVQGLLEIQHLTEKSLYNQRLRLQNEHRVLRQALRQKHQEAQQACRPHNLPVLQAAQQRELEAVEHRIREEQRAMDQKIVLELDRKVADQQSTLEKAGVAGFYVTTNPQELMLQMNLLELIRKLQQRGCWAGKAALGLGGPWQLPAAQCDQKGSPVPP

Specifications

Antigen DGCR6
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human DGCR6 protein.
Formulation PBS with no preservative; pH 7.4
Gene DGCR6
Gene Accession No. NM_005675.3
Gene Symbols DGCR6
Host Species Mouse
Immunogen DGCR6 (NP_005666.2, 1 a.a. ∼ 220 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8214
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.