Learn More
Description
Includes
Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-DGKA (100μg); Detection antibody: mouse purified polyclonal anti-DGKA (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human DGKA
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifications
Specifications
| Antigen | DGKA |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human DGKA. |
| Gene | DGKA |
| Gene Alias | DAGK/DAGK1/DGK-alpha/MGC12821/MGC42356 |
| Gene Symbols | DGKA |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.