Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disialoganglioside GD2 Mouse anti-Human, PE, Clone: 14.G2a, BD

Catalog No. BDB562100 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562100 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562100 Supplier BD Biosciences Supplier No. 562100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Gangliosides are sialic-acid bearing glycolipids that are expressed on the surface of all mammalian cells, and are likely involved in mediating cell-substratum interactions. They are important target antigens for antibody dependent cellular cytotoxicity (ADCC) of human melanoma and neuroblastoma cells. Human melanoma cells produce gangliosides, designated as GD2 and GD3 which are deposited in the subtratum-attached material, and may play a significant role in the melanoma metastatic phenotype. Clone 14.G2a specifically reacts with human and mouse GD2 ganglioside. LAN-1 human neuroblastoma cells were used as immunogen. Clone 14.G2a is an isotype switch variant selected from the parental IgG3-producing hybridoma 14.18 and has identical reactivity as the parental antibody. Clone 14.G2a is routinely tested by flow cytometry using M21 human melanoma cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Disialoganglioside GD2
Applications Flow Cytometry
Classification Monoclonal
Clone 14.G2a
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen LAN-1 human neuroblastoma cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.