Learn More
AdipoGen ANTI-DITYROSINE (DT), MAB (1C3
SDP
Supplier:  AdipoGen JAIMDT020P
Encompass_Preferred
anti-Dityrosine [DT], mAb (1C3), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes free dityrosine, 3-(p-hydroxyphenyl) propionic acid dimer, dityrosine-BSA conjugate, and dityrosine in protein or peptides. Does not cross-react with nitrotyrosine, chlorotyrosine, aminotyrosine, phosphotyrosine, tyrosine, 3-(p-hydroxyphenyl) propionic acid, phenylalanine, 3,4-dihydroxyphenylacetic acid (DOPA). Lyophilized. Contains 10mM PBS (pH7.4). Tyrosine is one of the major targets of protein oxidation. Dityrosine (DT) is known to be formed when tyrosine is damaged by free radicals, such as reactive oxygen species (ROS), metal-catalyzed oxidation, ultraviolet irradiation, and peroxidases. Dityrosine has been found in atherosclerotic lesions, and lipofuscin of pyramidal neurons of aged human brains.
Catalog No. NC0635294
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.