Learn More
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-DNAJB6 (100μg); Mouse purified polyclonal anti-DNAJB6 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
This gene encodes a member of the DNAJ protein family. DNAJ family members are characterized by a highly conserved amino acid stretch called the 'J-domain' and function as one of the two major classes of molecular chaperones involved in a wide range of cellular events, such as protein folding and oligomeric protein complex assembly. This family member may also play a role in polyglutamine aggregation in specific neurons. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been fully described.
ELISA
Specifications
Specifications
| Antigen | DNAJB6 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human DNAJB6. |
| Gene | DNAJB6 |
| Gene Alias | DJ4/DKFZp566D0824/DnaJ/FLJ42837/HHDJ1/HSJ-2/HSJ2/MGC1152/MGC117297/MRJ/MSJ-1 |
| Gene Symbols | DNAJB6 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.