Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doublecortin Mouse, PE, Clone: 30/Doublecortin, BD

Catalog No. BDB561505 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561505 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB561505 Supplier BD Biosciences Supplier No. 561505
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The cerebral cortex is composed of multiple layers of neurons of distinct types and functions, which perform in unison to orchestrate cognitive function. The formation of the cortex relies on complex signaling mechanisms, which mediate the migration of newly formed neurons from deep within the brain to the superficial regions. Defects in neuronal migration and disruption of the multi-layered cortex are apparent in conditions such as X-linked lissencephaly (XLIS) and subcortical laminar heterotopia (SCLH) or “double cortex” (DC) syndrome. Mutations in the doublecortin gene have been linked to these brain disorders. Doublecortin is highly expressed in developing brain, primarily in migrating neurons. It is significantly homologous with the N-terminal region of the product of the KIAA0369 gene, a protein that is also similar to the CaM kinase family in its C-terminal region. Doublecortin has four potential MAP kinase family phosphorylation sites and a putative site for Abl tyrosine phosphorylation. Thus, doublecortin is thought to be an integral component of tyrosine kinase signal transduction pathways that regulate neuronal migration and development of the cerebral cortex.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Doublecortin
Applications Flow Cytometry
Classification Monoclonal
Clone 30/Doublecortin
Conjugate PE
Description DCX, Lissencephalin-X, Lis-X, Doublin, Doublecortex, DBCN, LISX
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Doublecortin aa. 211-317 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.