Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSCR1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89003262 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-262 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-262 Supplier Abnova Corporation Supplier No. H00001827A02
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant DSCR1.

The protein encoded by this gene interacts with calcineurin A and inhibits calcineurin-dependent signaling pathways, possibly affecting central nervous system development. This gene is located in the minimal candidate region for the Down syndrome phenotype, and is overexpressed in the brain of Down syndrome fetuses. Chronic overexpression of this gene may lead to neurofibrillary tangles such as those associated with Alzheimer disease. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq

Sequence: MEEVDLRDLPSATIACHLDPRVFVDGLCRAKFESLFRTYDKDITFQYFKSFKRVRINFSNPFSAADARLQLHKTEFLGKEMKLYFAQTLHIGSSHLAPPNPDKQFLISPPASPPVGWKQVEDATPVINYDLLYAISKLGPGEKYELHAATDTTPSVVVHVCESDQEKEEEEEMERMRRPKPKIIQTRRPEYTQIHLS

Specifications

Antigen DSCR1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant DSCR1.
Formulation 50% glycerol
Gene RCAN1
Gene Accession No. BC002864
Gene Alias ADAPT78/CSP1/DSC1/DSCR1/MCIP1/RCN1
Gene Symbols RCAN1
Host Species Mouse
Immunogen DSCR1 (AAH02864, 1 a.a. ∼ 197 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1827
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.