Learn More
Clontech Labs 3P ANTI-DSRED MONOCLONAL ANTIBODY
Supplier: Clontech Labs 3P 632392
A monoclonal antibody for the detection of Discosoma sp. red fluorescent protein (DsRed). This antibody recognizes denatured forms of wild-type DsRed and its variants, including DsRed1, DsRed2, and DsRed1-E5 (the Fluorescent Timer).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.