Learn More
E-Cadherin Mouse, Alexa Fluor 555, Clone: 36/E-Cadherin, BD
Catalog No. BDB560064 Shop All BD Biosciences Products
Change view
Click to view available options
100 Tests; Alexa Fluor 555
Catalog No.
BDB560064 100 Tests; Alexa Fluor 555
1 options
This item is not returnable. View return policy
Catalog No. BDB560064 Supplier BD Biosciences Supplier No. 560064
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mouse Monoclonal Antibody
E-Cadherin is a 120-kDa transmembrane glycoprotein that is localized in the adherens junctions of epithelial cells. There it interacts with the cytoskeleton through the associated cytoplasmic catenin proteins. In addition to being a calcium-dependent adhesion molecule, E-Cadherin is also a critical regulator of epithelial junction formation. Its association with catenins is necessary for cell-cell adhesion. These E-cadherin/catenin complexes associate with cortical actin bundles at both the zonula adherens and the lateral adhesion plaques. Tyrosine phosphorylation can disrupt these complexes, leading to changes in cell adhesion properties. E-Cadherin expression is often down-regulated in highly invasive, poorly differentiated carcinomas. Increased expression of E-Cadherin in these cells reduces invasiveness. Thus, loss of expression or function of E-Cadherin appears to be an important step in tumorigenic progression. The 36/E-Cadherin monoclonal antibody recognizes the cytoplasmic domain of E-Cadherin, regardless of phosphorylation status. The peptide immunogen was generated from human E-Cadherin aa. 735-883.
Note: Investigators are advised that this antibody has some degree of cross-reactivity to P-Cadherin.
TRUSTED_SUSTAINABILITY
WARNING: Reproductive Harm - www.P65Warnings.ca.gov