Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFP Mouse anti-Human, Unlabeled, Clone: 2, BD

Catalog No. BDB610570 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610570 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610570 Supplier BD Biosciences Supplier No. 610570
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Steriod hormones, unlike polypeptide hormones, diffuse inside the cell, bind cytoplasmic or nuclear receptors, and activate specific genes. Complete identification of such receptors would greatly increase the understanding of the celluar processes activated by steriods. EFP (Estrogen receptor Finger Protein) was identified as a result of its specific interaction with the estrogen receptor. EFP contains a zinc-finger domain, indicating that it is a DNA-binding protein. EFP transcription is regulated by estrogens and, in combination with the estrogen receptor, it may modulate the expression of estrogen-responsive genes.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen EFP
Applications Western Blot
Classification Monoclonal
Clone 2
Concentration 250μg/mL
Conjugate Unconjugated
Description Estrogen receptor Finger Protein
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.