Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGR1/EGR2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA538588
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538588 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538588 Supplier Thermo Scientific Supplier No. PA538588

Rabbit Polyclonal Antibody

The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGR1/EGR2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EGR1
Gene Accession No. P11161, P18146, P08046, P08152, P08154, P51774
Gene Alias TIS8, AT225, G0S30, NGFI-A, ZNF225, KROX-24, ZIF-268, EGR1, EGR2; early growth response 2 (Krox-20 homolog, Drosophila) ; CMT1D; CMT4E; DKFZp686J1957; FLJ14547; KROX20; KROX-20, Drosophila, homolog early growth response-2; Krox-20 homolog
Gene Symbols EGR1, EGR2
Host Species Rabbit
Immunogen A synthetic peptide derived from the internal region of human EGR1/2
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114090, 13653, 13654, 1958, 1959, 24330
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.