Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eukaryotic translation initiation factor 2C, 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004812 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-812 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-812 Supplier Abnova Corporation Supplier No. H00192669A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant EIF2C3.

This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic, contains a PAZ domain and a PIWI domain, and may play a role in short-interfering-RNA-mediated gene silencing. This gene is located on chromosome 1 in a tandem cluster of closely related family members including argonaute 4 and eukaryotic translation initiation factor 2C, 1. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Sequence: MEIGSAGPAGAQPLLMVPRRPGYGTMGKPIKLLANCFQVEIPKIDVYLYEVDIKPDKCPRRVNREVVDSMVQHFKVTIFGDRRPVYDGKRSLYTANPLPV

Specifications

Antigen eukaryotic translation initiation factor 2C, 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant EIF2C3.
Formulation 50% glycerol
Gene EIF2C3
Gene Accession No. NM_024852
Gene Alias AGO3/FLJ12765/MGC86946
Gene Symbols EIF2C3
Host Species Mouse
Immunogen EIF2C3 (NP_079128, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 192669
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.