Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

erythrocyte membrane protein band 4.1 like 4A, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008052 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-052 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-052 Supplier Abnova Corporation Supplier No. H00064097B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human EPB41L4A protein.

Members of the band 4.1 protein superfamily, including EPB41L4A, are thought to regulate the interaction between the cytoskeleton and plasma membrane (Ishiguro et al., 2000 [PubMed 10874211]).[supplied by OMIM

Sequence: MGCFCAVPEEFYCEVLLLDESKLTLTTQQQGIKKSTKGSVVLDHVFHHVNLVEIDYFGLRYCDRSHQTYWLDPAKTLAEHKELINTGPPYTLYFGIKFYAEDPCKLKEEITRYQFFLQVKQDVLQGRLPCPVNTAAQLGAYAIQSELGDYDPYKHTAGYVSEYRFVPDQKEELEEAIERIHKTLM

Specifications

Antigen erythrocyte membrane protein band 4.1 like 4A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human EPB41L4A protein.
Formulation No additive
Gene EPB41L4A
Gene Accession No. BC031042.1
Gene Alias EPB41L4/FLJ38738/NBL4
Gene Symbols EPB41L4A
Host Species Mouse
Immunogen EPB41L4A (AAH31042.1, 1 a.a. ∼ 185 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 64097
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.