Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA4/Sek Mouse, Unlabeled, Clone: 35, BD

Catalog No. BDB610471 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610471 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610471 Supplier BD Biosciences Supplier No. 610471
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

EphA4/Sek is a glycosylated protein tyrosine kinase of 986 amino acids and an apparent 110-130kDa molecular weight (molecular weight depends on glycocylation level of protein). EphA4 belongs to the Eph subfamily of receptor tyrosine kinases and like many others, it has an amino-terminal ligand-binding domain exposed to the extracellar matrix. EphA4 is predominantly expressed during development (this is common in the Eph family) and because of its mRNA localization, it has been proposed that it may play a role during segmentation of the hindbrain and mesoderm. Tyrosine 602 is a major autophosphorylation site of EphA4 and appears to be the docking site for the intracellular tyrosine kinase fyn.

TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA4/Sek
Applications Western Blot
Classification Monoclonal
Clone 35
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.