Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eps15 Mouse, Unlabeled, Clone: 17, BD

Catalog No. BDB610806 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610806 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610806 Supplier BD Biosciences Supplier No. 610806
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The eps15 (EGF receptor pathway substrate clone number 15) gene was isolated using an expression cloning method designed for direct isolation of cDNAs encoding substrates of tyrosine kinases. The eps15 gene encodes a 145kDa protein that is tyrosine phosphorylated following EGF receptor activation and this receptor directly phosphorylates purified Eps15 in vitro. Phosphorylation of Eps15 is much more efficient following EGF receptor activation versus erbB2 kinase activation, suggesting the protein is predominantly a part of an EGF receptor signaling pathway. Overexpression of Eps15 causes transformation of NIH 3T3 cells, implying Eps15 may be involved in the control of cell proliferation. Eps15 binds to the SH3 domain of Crk through its proline-rich domain at the C-terminal region. Adaptin α binds to the C-terminus of Eps15 at three different sites ranging from residues 650 to 730. Eps15, Adaptin α and EGF receptor are co-localized at the clathrin-coated pits after receptor activation. Therefore, Eps15 may recruit EGF receptors to the clathrin-coated pits for internalization of the activated receptor.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Eps15
Applications Western Blot
Classification Monoclonal
Clone 17
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Eps15 aa. 694-888
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.