Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ERBB3 & ERBB2 Protein Protein Interaction Antibody Pair, Abnova™

Catalog No. 89023832 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-023-832 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-023-832 Supplier Abnova Corporation Supplier No. DI0159

Immunofluorescence

Specifications

Antigen ERBB3
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene ERBB3
Gene Alias ErbB-3/HER3/LCCS2/MDA-BF-1/MGC88033/c-erbB-3/c-erbB3/erbB3-S/p180-ErbB3/p45-sErbB3/p85-sErbB3
Gene Symbols ERBB3
Quantity 1 Set
Primary or Secondary Primary
Gene ID (Entrez) 2065/2064
Includes Antibody pair set content: 1. ERBB3 rabbit purified polyclonal antibody (20 ug) 2. ERBB2 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type ERBB3 & ERBB2 Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.