Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrogen Receptor Β Rabbit anti-Human, Clone: 68-4, MilliporeSigma™

Catalog No. 05824MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-824-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-824-MI Supplier MilliporeSigma Supplier No. 05824
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

Detect Estrogen Receptor β also known as Nuclear receptor subfamily 3 group A member 2 with Anti-Estrogen Receptor β Antibody, clone 68-4 (Rabbit Monoclonal Antibody), that has been demonstrated to work in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Estrogen Receptor β
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 68-4
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl and 0.05% sodium azide, pH 7.4.
Gene Accession No. NM_001437
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 63-82 of rat and mouse estrogen receptor β and amino acids 55-74 of human estrogen receptor β
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) ESR2, ESR-BETA, ESTRB, ESRB, ER-BETA, NR3A2, Erb, ER-beta
Target Species Human
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.