Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-F4/80, Clone: BM8, eBioscience™

Catalog No. NC0066541
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC0066541 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC0066541 Supplier Biolegend Supplier No. 123113
Only null left
Add to Cart
Add to Cart

Specifications

Antigen F4/80
Classification Monoclonal
Clone BM8
Concentration 0.2mg/mL
Conjugate PE-Cyanine7
Host Species Rat
Purification Method Unpurified
Quantity 25 μg
Primary or Secondary Primary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.