Learn More
Abcam Anti-Fade Fluorescence Mounting Medium, Aqueous, Fluoroshield, Liquid, 100 mL
Description
- Aqueous mounting medium designed to preserve fluorescence in tissue and cell samples
- Prevents rapid photobleaching of key fluorophores
- Contains 84% water
- Preservative: 0.05% Sodium azide
- Fluorescence remains stable at +4°C in the dark
- Cited in over 30 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Content And Storage | Store at 4°C in the dark. |
| Form | Liquid |
| Detection Method | Fluorescence |
| Buffer | 0.05% sodium azide, 84% water |
| Formulation | 84% water, 0.05% sodium azide |
| Sample Type | Tissues, Cell samples |
| Quantity | 100 mL |
| For Use With (Application) | Fluorescence imaging of tissues and cell samples |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.