Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAK (pS910) Mouse, PE, Clone: K73-480, BD

Catalog No. BDB558540 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558540 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558540 Supplier BD Biosciences Supplier No. 558540
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Focal Adhesion Kinase (FAK) is a cytoplasmic tyrosine kinase that associates with integrins in focal adhesions. Its cellular localization is directed by a 125-amino acid sequence at the C terminus called the “Focal Adhesion Targeting” (FAT) domain, and the phosphorylation state of serine 910 (S910) in the FAT domain may regulate the assembly of focal adhesions. Furthermore, the binding of extracellular matrix ligands to integrins triggers tyrosine phosphorylations near FAK's kinase domain that increase its kinase activity, and additional tyrosine phosphorylations near proline-rich motifs create binding sites for the SH2 domains of various adaptor proteins. FAK's ability to bind numerous structural and signaling proteins via a variety of interactions regulates FAK's targeting to focal adhesions, modulates its kinase activity, and initiates intracellular signaling cascades. Thus, studies suggest that FAK may integrate cellular events controlling cell motility, growth, and invasiveness. The K73-480 monoclonal antibody recognizes the phosphorylated S910 of human FAK. The orthologous phosphorylation sites in mouse and rat FAK are S948 and S913, respectively.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAK (pS910)
Applications Flow Cytometry
Classification Monoclonal
Clone K73-480
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human FAK Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.