Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FASLG, FN1 Protein Protein Interaction Antibody Pair, Abnova™

Catalog No. 89003525 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-525 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-525 Supplier Abnova Corporation Supplier No. DI0606
Add to Cart
Add to Cart

Includes

FASLG rabbit purified polyclonal antibody (20μg); FN1 mouse purified polyclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols.

Host Species: Rabbit, Mouse
Cross Reactivity: Human

In situ PLA

Specifications

Antigen FASLG
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene FASLG
Gene Alias APT1LG1/CD178/CD95L/FASL/TNFSF6
Gene Symbols FASLG
Host Species Mouse, Rabbit
Quantity 1 Set
Primary or Secondary Primary
Gene ID (Entrez) 356/2335
Includes Antibody pair set content: 1. FASLG rabbit purified polyclonal antibody (20 ug) 2. FN1 mouse purified polyclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type FASLG & FN1 Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.