Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAT10 Mouse anti-Human, Mouse, Rat, Clone: 4F1, MilliporeSigma™

Catalog No. MABS351MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS351MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS351MI Supplier MilliporeSigma Supplier No. MABS351
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-FAT10 Antibody, clone 4F1 is a highly specific mouse monoclonal antibody, that targets FAT10 and has been tested in western blotting, immunoprecipitation and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAT10
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4F1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2a in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_006389
Gene Symbols UBD, FAT10
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human FAT10.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.