Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fc gamma RIII (CD16) Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$152.84 - $342.67
Specifications
| Antigen | Fc gamma RIII (CD16) |
|---|---|
| Clone | 245536 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB2546F025
![]() |
R&D Systems
FAB2546F025 |
25 Tests |
Each for $152.84
|
|
|||||
FAB2546F100
![]() |
R&D Systems
FAB2546F100 |
100 Tests |
Each for $342.67
|
|
|||||
Description
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Flow Cytometry.Specifications
| Fc gamma RIII (CD16) | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD16, CD16A, CD16b, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGR3A, FCGR3B, FCGRIII, FcgRIIIA, FcgRIIIB, FcR-10, FCRIII, FCRIIIA, FCRIIIB, IGFR3, IMD20 | |
| FCGR3A | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human Fc gamma RIIIA/B (CD16) in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Fc gamma RIIA and recombinant mouse Fc gamma RIII is observed. |
| 245536 | |
| Flow Cytometry | |
| Fluorescein | |
| Mouse | |
| Human | |
| O75015 | |
| 2214 | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIIA/B (CD16) Thr20-Gln208 Accession # O75015 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title