Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Anti-Ferritin Antibody [RM1036], Rabbit Multiclonal, 100 μL

Catalog No. 502617434
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-261-7434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-261-7434 Supplier Abcam Supplier No. AB287968100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit recombinant multiclonal antibody for Western blot, immunohistochemistry, and intracellular flow cytometry applications.

  • Host Species: Rabbit
  • Clonality: Multiclonal
  • Isotype: IgG
  • Applications: WB, IHC-P, Flow Cyt (Intra)
  • Species Reactivity: Human, Mouse, Rat
  • Form: Liquid
  • Storage Buffer: PBS, 40% Glycerol, 0.05% BSA, 0.01% Sodium azide, pH 7.2 to 7.4
  • Citations: 8 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Ferritin Heavy Chain (Human, Mouse, Rat)
Label or Dye Unconjugated
Detection Location In-Blot Detection
Buffer PBS, 40% glycerol, 0.05% BSA, 0.01% sodium azide, pH7.2 to 7.4
Form Liquid
Cross Reactivity Mouse, Human, Rat
Formulation PBS, 40% glycerol, 0.05% BSA, 0.01% sodium azide, pH 7.2 to 7.4
Quantity 100 μL
For Use With (Application) Flow Cytometry (Intra), Western Blot, IHC-P
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.