Learn More
Description
Includes
Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FGD3 (100μg); Detection antibody: mouse purified polyclonal anti-FGD3 (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Host Species: Rabbit, Mouse
Cross Reactivity: Human
ELISA Pair
Specifications
Specifications
| Antigen | FGD3 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FGD3. |
| Gene | FGD3 |
| Gene Alias | FLJ00004/MGC117260/ZFYVE5 |
| Gene Symbols | FGD3 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.