Learn More
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-FGF21 (100μg); Biotinylated rabbit MaxPab™ affinity purified polyclonal anti-FGF21 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
ELISA
Specifications
Specifications
| Antigen | FGF21 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Biotin |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FGF21. |
| Gene | FGF21 |
| Gene Symbols | FGF21 |
| Host Species | Rabbit |
| Purification Method | Affinity Purified |
| Quantity | 1 Set |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.