Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP12 Mouse, Unlabeled, Clone: 8, BD

Catalog No. BDB610808 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610808 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610808 Supplier BD Biosciences Supplier No. 610808
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transforming growth factor-β binds to the TGFβ family of heteromeric serine/threonine transmembrane receptors (type I and type II). Following binding of TGFβ, the type II receptor (TβR-II) phosphorylates the type I receptor (TβR-I) which, in turn, conveys the signal. Since TβR-I and TβR-II can interact without the stimulation of TGFβ, leading to unwanted activation, a regulatory mechanism exists. In a yeast genetic screen, immunophilin FKBP12 was associated with the type I receptor. Studies including co-immunoprecipiation, deletion, and point mutations confirmed this interaction. FKBP12 inhibits TβR-II mediated phosphorylation of TβR-I, inhibiting activation. FKBP12 binds via its rapamycin/Leu-Pro binding pocket to the Leu-Pro sequence adjacent to the phosphorylation site of TβR-I. This interaction is blocked by
the addition of macrolides, rapamycin, and FK506. Furthermore, mutations in the binding sites of FKBP12 and TβR-I abrogates the binding and results in activation of the receptor without the addition of TGFβ. Thus, FKBP12 is a regulatory protein for TβR-I and TβR-II-mediated signaling.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP12
Applications Western Blot
Classification Monoclonal
Clone 8
Concentration 250μg/mL
Conjugate Unconjugated
Description FK506 Binding Protein 1A
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human FKBP12 aa. 1-108
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.